Anti-LRP1 Mouse Monoclonal Antibody [clone: 5A6]

Supplier: Innovative Research Inc
MA-5A6-10MG MA-5A6-1MG
103387-106EA 4947.69 USD
103387-106 103388-064
Anti-LRP1 Mouse Monoclonal Antibody [clone: 5A6]
Antibodies
Type: Primary
Antigen: LRP1
Clonality: Monoclonal
Clone: 5A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human;Mouse;Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR