Anti-VLDLR Mouse Monoclonal Antibody [clone: 1H5]

Supplier: Innovative Research Inc
MA-1H5-10MG MA-1H5-1MG
103387-090EA 4947.69 USD
103387-090 103388-048
Anti-VLDLR Mouse Monoclonal Antibody [clone: 1H5]
Antibodies
Type: Primary
Antigen: VLDLR
Clonality: Monoclonal
Clone: 1H5
Conjugation: Unconjugated
Epitope: C-terminus
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR