Anti-KNG1 Mouse Monoclonal Antibody [clone: 17E10-G9]

Supplier: Innovative Research Inc
KNG17E10-0.1MG KNG17E10-1MG
103385-402EA 246.15 USD
103385-402 103387-072
Anti-KNG1 Mouse Monoclonal Antibody [clone: 17E10-G9]
Antibodies
Type: Primary
Antigen: KNG1
Clonality: Monoclonal
Clone: 17E10-G9
Conjugation: Unconjugated
Epitope: Light Chain
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR