Anti-KNG1 Mouse Monoclonal Antibody [clone: 15G5-A11]

Supplier: Innovative Research Inc
KNG15G5-0.1MG KNG15G5-1MG
103385-396EA 246.15 USD
103385-396 103387-066
Anti-KNG1 Mouse Monoclonal Antibody [clone: 15G5-A11]
Antibodies
Type: Primary
Antigen: KNG1
Clonality: Monoclonal
Clone: 15G5-A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR