Anti-PLAU Mouse Monoclonal Antibody [clone: 724-3G6-5] (FITC (Fluorescein Isothiocyanate))

Supplier: Innovative Research Inc
HUPA3G65FITC.1 HUPA3G65FITC1
103385-350EA 375.38 USD
103385-350 103388-690
Anti-PLAU Mouse Monoclonal Antibody [clone: 724-3G6-5] (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Primary
Antigen: PLAU
Clonality: Monoclonal
Clone: 724-3G6-5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: beta chain of Urokinase (uPA)
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR