Anti-PLAU Mouse Monoclonal Antibody [clone: 724-3G6-5]

Supplier: Innovative Research Inc
HUPA3G65-1MG HUPA3G65-10MG
103385-348EA 547.69 USD
103385-348 103388-350
Anti-PLAU Mouse Monoclonal Antibody [clone: 724-3G6-5]
Antibodies
Type: Primary
Antigen: PLAU
Clonality: Monoclonal
Clone: 724-3G6-5
Conjugation: Unconjugated
Epitope: beta chain of Urokinase (uPA)
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR