Anti-IGHG3 Human Antibody

Supplier: Innovative Research Inc
HU-IGG3-1MG HU-IGG3-5MG
103385-330EA 613.85 USD
103385-330 103387-024
Anti-IGHG3 Human Antibody
Antibodies
Type: Control
Antigen: IGHG3
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR