Anti-IGHA1 Human Antibody

Supplier: Innovative Research Inc
HU-IGA-5MG HU-IGA-10MG
103385-304EA 204.62 USD
103385-304 103386-998
Anti-IGHA1 Human Antibody
Antibodies
Type: Control
Antigen: IGHA1
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgA
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR