Anti-PLAU Mouse Monoclonal Antibody [clone: H77B6]

Supplier: Innovative Research Inc
H77B6-1MG H77B6-10MG
103384-986EA 869.62 USD
103384-986 103386-728
Anti-PLAU Mouse Monoclonal Antibody [clone: H77B6]
Antibodies
Type: Primary
Antigen: PLAU
Clonality: Monoclonal
Clone: H77B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR