Anti-SERPINE1 Mouse Monoclonal Antibody [clone: H4B3]

Supplier: Innovative Research Inc
H4B3-1MG H4B3-10MG
103384-980EA 869.62 USD
103384-980 103386-722
Anti-SERPINE1 Mouse Monoclonal Antibody [clone: H4B3]
Antibodies
Type: Primary
Antigen: SERPINE1
Clonality: Monoclonal
Clone: H4B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse;Rat;Human;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR