Anti-SERPINE1 Mouse Monoclonal Antibody [clone: H34G6]

Supplier: Innovative Research Inc
H34G6-1MG H34G6-10MG
103384-978EA 869.62 USD
103384-978 103386-720
Anti-SERPINE1 Mouse Monoclonal Antibody [clone: H34G6]
Antibodies
Type: Primary
Antigen: SERPINE1
Clonality: Monoclonal
Clone: H34G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse;Rat;Rabbit;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR