Anti-SERPINE1 Mouse Monoclonal Antibody [clone: H14H7] (Biotin)

Supplier: Innovative Research Inc
H14H7BIO0.15MG H14H7BIO1.05MG
103384-974EA 825.77 USD
103384-974 103386-716
Anti-SERPINE1 Mouse Monoclonal Antibody [clone: H14H7] (Biotin)
Antibodies
Type: Primary
Antigen: SERPINE1
Clonality: Monoclonal
Clone: H14H7
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse;Rat;Human;Rabbit;Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR