Anti-IgG Goat Polyclonal Antibody

Supplier: Innovative Research Inc
GT-GF-50MG GT-GF-100MG
103384-970EA 104.62 USD
103384-970 103386-712
Anti-IgG Goat Polyclonal Antibody
Antibodies
Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR