Anti-F7 Mouse Monoclonal Antibody [clone: 11G42-D8] (Biotin)

Supplier: Innovative Research Inc
FVII11G42BIO10 FVII11G42BIO1
103386-626EA 5618.46 USD
103386-626 103388-000
Anti-F7 Mouse Monoclonal Antibody [clone: 11G42-D8] (Biotin)
Antibodies
Type: Primary
Antigen: F7
Clonality: Monoclonal
Clone: 11G42-D8
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR