Anti-F5 Mouse Monoclonal Antibody [clone: 14G5-G11] (Biotin)

Supplier: Innovative Research Inc
FV14G5BIO0.1MG FV14G5-BIO-1MG
103384-912EA 375.38 USD
103384-912 103386-622
Anti-F5 Mouse Monoclonal Antibody [clone: 14G5-G11] (Biotin)
Antibodies
Type: Primary
Antigen: F5
Clonality: Monoclonal
Clone: 14G5-G11
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR