Anti-ALB Mouse Monoclonal Antibody [clone: 12C8-D4]

Supplier: Innovative Research Inc
BSA12C8D4-1MG BSA12C8D4-10MG
103384-772EA 276.92 USD
103384-772 103386-514
Anti-ALB Mouse Monoclonal Antibody [clone: 12C8-D4]
Antibodies
Type: Primary
Antigen: ALB
Clonality: Monoclonal
Clone: 12C8-D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR