Anti-SERPINE1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Innovative Research Inc
ASRPAIGFBIO1MG ASRPAIBIO10MG
103384-744EA 498.46 USD
103384-744 103386-486
Anti-SERPINE1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: SERPINE1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR