Anti-VTN Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Innovative Research Inc
ASMVNGFHRP1MG ASMVNGFHRP10MG
103384-724EA 578.46 USD
103384-724 103388-278
Anti-VTN Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Primary
Antigen: VTN
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR