Anti-PLAU Rabbit Polyclonal Antibody

Supplier: Innovative Research Inc
ASMUPA-1ML ASMUPA-10ML
103384-712EA 375.38 USD
103384-712 103388-620
Anti-PLAU Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PLAU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR