Anti-F12 Rabbit Polyclonal Antibody (Biotin)

Supplier: Innovative Research Inc
ASMFXIIHTBIO1 ASMFXIIHTBIO.1
103386-426EA 3987.69 USD
103386-426 103388-846
Anti-F12 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: F12
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR