Anti-F12 Rabbit Polyclonal Antibody

Supplier: Innovative Research Inc
ASMFXIIGFHT1MG ASMFXIIHT0.1MG
103386-424EA 3483.08 USD
103386-424 103388-844
Anti-F12 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: F12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR