Anti-F12 Rabbit Polyclonal Antibody

Supplier: Innovative Research Inc
ASMFXII-GF-1MG ASMFXIIGF10MG
103384-674EA 572.31 USD
103384-674 103386-416
Anti-F12 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: F12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR