Anti-FN1 Rabbit Polyclonal Antibody

Supplier: Innovative Research Inc
ASMFBN-10ML ASMFBN-1ML
103386-402EA 2978.46 USD
103386-402 103387-984
Anti-FN1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR