Anti-FGA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Innovative Research Inc
ASMFBGNHRP10MG ASMFBGNHRP1MG
103386-398EA 3987.69 USD
103386-398 103387-980
Anti-FGA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Primary
Antigen: FGA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR