Anti-VTN Rabbit Polyclonal Antibody (Biotin)

Supplier: Innovative Research Inc
ASHVNGFBIO10MG IRBAHUVTNGFBC1MG
103386-370EA 4596.92 USD
103386-370 103387-936
Anti-VTN Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: VTN
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR