Anti-PLAT Rabbit Polyclonal Antibody (Biotin)

Supplier: Innovative Research Inc
ASHTPABIO10MG ASHTPAGFBIO1MG
103386-344EA 4024.62 USD
103386-344 103387-918
Anti-PLAT Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: PLAT
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR