Anti-PLAT Rabbit Polyclonal Antibody

Supplier: Innovative Research Inc
ASHTPA-10ML ASHTPA-1ML
103386-340EA 3636.92 USD
103386-340 103387-914
Anti-PLAT Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PLAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR