Anti-REN Rabbit Polyclonal Antibody

Supplier: Innovative Research Inc
ASHPRENGF10MG ASHPREN-GF-1MG
103386-318EA 3280 USD
103386-318 103388-818
Anti-REN Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: REN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR