Anti-SERPINE1 Rabbit Polyclonal Antibody

Supplier: Innovative Research Inc
ASHPAI-GF-10MG ASHPAI-GF-1MG
103386-304EA 3636.92 USD
103386-304 103387-894
Anti-SERPINE1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SERPINE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR