Anti-F10 Rabbit Polyclonal Antibody (Biotin)

Supplier: Innovative Research Inc
ASHFXGFBIO10MG ASHFXGFBIO1MG
103386-288EA 3840 USD
103386-288 103387-878
Anti-F10 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: F10
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR