Anti-F10 Rabbit Polyclonal Antibody

Supplier: Innovative Research Inc
ASHFX-10ML ASHFX-1ML
103386-284EA 3280 USD
103386-284 103387-874
Anti-F10 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: F10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR