Anti-FGA Rabbit Polyclonal Antibody

Supplier: Innovative Research Inc
ASHFBGN-GF-1MG ASHFBGNGF10MG
103384-644EA 430.77 USD
103384-644 103386-250
Anti-FGA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FGA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR