Anti-SERPINF2 Rabbit Polyclonal Antibody

Supplier: Innovative Research Inc
ASHA2APHT0.1MG ASHA2APGFHT1MG
103384-582EA 443.08 USD
103384-582 103386-220
Anti-SERPINF2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SERPINF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR