Digital Dispensers, D300e, Dispensehead Cassettes, Tecan
Supplier: TECAN US INC MS
30097370 30097371
MSPP-30097370PK 2783.17 USD
MSPP-30097370 MSPP-30097371
Digital Dispensers, D300e, Dispensehead Cassettes, Tecan
Microplate Dispensers
The T8+ and D4+ Dispensehead cassettes allow dispensing of up to eight fluids per cassette, from 11 picoliters to 10 microliters of small molecules and biomolecules in DMSO or aqueous solution.
  • Plate formats: 12 to 1536 well plates, 6 to 47 mm plate height
  • T8+ can be also used for titration and low volume dispensing