Poly(diallyldimethylammonium chloride) [pDADMAC] 10% in water flocculation reagent, Millipore®

Supplier: MilliporeSigma

Synonyms: Polyquaternium-6, N,N-Dimethyl-N-2-propenyl-2-propen-1-aminiumchloridehomopolymer, PolyDADMAC

1.37069.1000 1.37069.9010
EM1.37069.1000EA 3631.03 USD
EM1.37069.1000 EM1.37069.9010
Poly(diallyldimethylammonium chloride) [pDADMAC] 10% in water flocculation reagent, Millipore®
Poly(diallyldimethylammonium chloride) [pDADMAC]

pDADMAC is a water-soluble polymer widely used as a flocculant in waste water treatment. The purification process is designed to meet GMP requirements. pDADMAC delivers the best flocculation results when used in a concentration between 0.01 and 0.05%. Since the solution is of neutral pH, only slight pH adjustments may be needed during use.


Sterile filtration steps reduce the microbial count to less than 10 CFU/ml.

Formula: (C₈H₁₆ClN)n
Density: 1.015 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00192409
CAS Number: 26062-79-3

Order Now

Specification Test Results

Identity (NIR) Passes Test
Appearance Colorless to Yelowish Liquid
Dry Residue (105 °C) 9.0 - 11.0%
Total Aerobic Microbial Count (TAMC) ≤10 CFU
Total Yeast and Mould Count (TYMC) ≤10 CFU
Bile Tolerant Gram Negative Bacteria Absent in 1 ml
Candida albicans Absent in 1 ml
E. coli Absent in 1 ml
Pseudomonas aeruginosa Absent in 1 ml
Salmonella Species Absent in 1 ml
Staphylococcus aureus Absent in 1 ml
Endotoxins ≤500 I.U/ml
Monomer Content ≤0.1%
Viscosity (20 °C) 100 - 350 mPa.s
Residual Solvents Excluded in Manufacturing Process

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR