DPD Liquid Reagents, LaMotte
Supplier: LA MOTTE CHEMICAL PRODUCTS CO
P6743-G P6741-H P6740-H P6743-H P-6740-G P6741-G
82013-848EA 18.74 USD
82013-848 82013-840 82013-838 82013-842 82013-844 82013-846
DPD Liquid Reagents, LaMotte
Environmental Test Kits
These reagents are liquid alternative to DPD tablets and can be used with existing chlorine comparators or colorimeters.
  • Used to measure pH, hardness, alkalinity, copper and other liquid reagents
DPD 1A and DPD 1B are added to test free available chlorine, DPD 3 added to the reacted sample to measure total chlorine.