Jackson Safety® G35 Inspection Gloves, Kimberly-Clark
Supplier: Ansell Healthcare
38716 38717 38718 38719 38720
10789-242BG 37.2 USD
10789-242 10789-244 10789-246 10789-248 10789-314
Jackson Safety® G35 Inspection Gloves, Kimberly-Clark
Gloves
Designed for conducting sensitive critical quality control inspections.
  • Low lint
  • 100% nylon knitted shell
  • Seamless construction
  • Silicone free
  • Excellent sensitivity and dexterity
The low lint 100% nylon material can help protect the inspection process and reduce rework in a way cotton gloves cannot. They feature a seamless construction and offer excellent dexterity and sensitivity so that inspectors can detect imperfections and defects.
The gloves are not made with silicone, so you can comfortably use them for quality inspections in a variety of environments, including film handling and the automotive and aerospace industries.