Chem-Ply N-440 Chemical Barrier Gloves, Mapa Professional
Supplier: MAPA
407959 407950
32929-136CS 1065.32 USD
32929-136 32929-138 32929-140
Chem-Ply N-440 Chemical Barrier Gloves, Mapa Professional
Gloves
These gloves are ultra-high performance chemical protection glove.
  • Broad chemical resistance
  • Embossed Z-pattern grip
  • Damage easily visible
  • Easy to pull on and off, due to chlorinated interior
The two neoprene layers superimposed increase the mechanical and chemical resistance.
The gloves red inner layer ensures that any wear is clearly visible, providing optimal safety for the user.
Certificaciones: CE Cat. III marked. Chemical Protection EN 374: ABCJKL. Mechanical Hazards EN 388: 3112.