Fast red TR salt (hemi zinc chloride salt) 10 - 12% (dye content)

Supplier: CHEM IMPEX INTL INC MS

00410-1KG 00410-100G 00410-250G
102606-416EA 281.92 USD
102606-416 102615-218 102615-220
Fast red TR salt (hemi zinc chloride salt) 10 - 12% (dye content)
Fast Red TR salt (hemi zinc chlroide salt)
Formula: C₁₄H₁₂Cl₆N₄Zn
Storage Temperature: Ambient
CAS Number: 89453-69-0

Order Now

Specification Test Results

Appearance Beige to Pinkish Solid
Cation (Zinc Content) 11.00 - 14.00%
Dye Content 10 - 12%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR