(S)-(-)-3-Acetylthio-2-methylpropionic acid ≥ ee 99% (by GC), liquid

Supplier: CHEM IMPEX INTL INC MS
Danger

05381-5G 05381-1G
102556-546EA 550.51 USD
102556-546 102619-562
(S)-(-)-3-Acetylthio-2-methylpropionic acid ≥ ee 99% (by GC), liquid
(S)-(-)-3-Acetylthio-2-methylpropionic acid

It can be used as an intermediate in the syntheses of cardiocasculars, in particular ACE-inhibitors.

Formula: C₆H₁₀O₃S
Density: 1,18
Flash Pt: >112 °C
Storage Temperature: Refrigerator
CAS Number: 76497-39-7
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Yellow Liquid
Specific Gravity 1.1770 - 1.1820 (20/20 °C)
Refractive Index (n20D) 1.4880 - 1.4900
Assay by Titration ≥99%
Chiral Purity ≥99% ee (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR