VHR Advanced Cut Protection Gloves, Portwest
Supplier: Portwest
A665GRRL A665GRRM A665GRRXL A665GRRXXL
76103-418EA 12.03 USD
76103-418 76103-420 76103-422 76103-424
VHR Advanced Cut Protection Gloves, Portwest
Gloves
VHR advanced cut protection gloves offer maximum EN cut protection levels.
  • Seamless 13 gauge liner
  • HDPE and glass fiber liner material
  • Palm dipped to increase dexterity and ventilation
The nitrile foam coating provides excellent grip in both wet and dry conditions.
Certificaciones: AS/NZS 2161 .2. AS/NZS 2161 .3. ANSI/ISEA 105 - 2016 CUT Level A6. EN 388:2003 - 4.5.4.4. EN 388:2016 - 4.X.4.4.E. EN 420. CE food safe. CE marked.