ActivArmr® 43-113 Heat resistant gloves, Ansell
Supplier: Ansell Healthcare
245638 243406
76020-750CS 1235.91 USD
76020-750 76020-768
ActivArmr® 43-113 Heat resistant gloves, Ansell
Gloves
Superior heat and flame resistance with complete protection for hand and wrist.
  • Resistant to contact, convective, and radiant heat
  • Flame resistant
  • Long knit wrist cuff extends protection to the wrist
  • Comfortable and absorbent inner cotton liner
Made with Dupont™ Kevlar® outer yarn, gloves are designed for intermittent handling of dry hot objects up to 350 °C (660 °F). The flame resistant material will not melt, drip or propagate a flame, providing the ultimate protection to the wearer.
Certificaciones: EN 407: 2004 - 43421X heat and fire protection.More Product Information
Order Now
SPECIFICATIONS
Model 43-113
Environmentally Preferable
Ambidextrous/Hand-specific Hand-Specific
Disposable/Reusable Reusable
Liner Material 100% Cotton
Material Kevlar®
Gauge 7
Cuff Style Knit Wrist