AlphaTec® 87-320 Lightweight Natural Rubber Gloves with Flocked Lining, Ansell
Supplier: Ansell Healthcare
87320075 87320085 87320095 87320065
76169-821CS 294.06 USD
76169-821 76169-823 76169-825 76169-875
AlphaTec® 87-320 Lightweight Natural Rubber Gloves with Flocked Lining, Ansell
Gloves
Gloves are comfortable, resistant and flexible. They are flock lined and light weight.
  • Beaded cuff for tear resistance and easy donning
  • Easy donning
  • Good abrasion resistance
  • Reversed lozenge grip design
  • Silicone free
  • 100% cotton flocked lining
For use in low hazard areas, primarily food processing applications.
Certificaciones: CE 0086 marked. EN ISO 374-1 Type B. EN374. EN ISO 374-5. EN388:2016 2010X. EN388:2003 2010.