Fmoc-L-cysteine hydrate ≥98% (by HPLC)
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
Fmoc-Cys-OH hydrate, N-(9H-Fluoren-9-ylmethoxycarbonyl)cysteine hydrate
14009-25G
14009-1G
14009-5G
102563-002EA
2190.85
USD
102563-002
102577-814
102594-212
Fmoc-L-cysteine hydrate ≥98% (by HPLC)
Fmoc-L-cysteine hydrate
Formula:
C₁₈H₁₇NO₄S·H₂O MW: 361.40 g/mol Melting Pt: 109…115 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD13184915 CAS Number: 135248-89-4 |
Specification Test Results
Appearance | White crystalline powder |
Melting Point | 109-115 °C |
[a]D20 (C=1 in DMF) | -21.0 to -26.0º |
[a]D20 (C=1 in MeOH) | -4.0 to -6.0º |
Purity by HPLC | ≥ 98% |
Purity by TLC | ≥ 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...