(S)-2-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)propanoic acid ≥99% (by HPLC), white powder
Supplier: CHEM IMPEX INTL INC MS
Synonyms:
Z-Dap(Boc)-OH, Nα-Z-Nβ-Boc-L-2,3-diaminopropionic acid, Z-3-(Boc-amino)-L-alanine, Cbz-Dap(Boc)-OH, 2-Benzyloxy-carbonyl-amino-3-tert-butoxycarbonyl-amino-propanoic-acid
04977-5G
04977-1G
04977-25G
102553-684EA
99.83
USD
102553-684
102556-450
102591-308
(S)-2-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)propanoic acid ≥99% (by HPLC), white powder
(S)-2-(((Benzyloxy)carbonyl)amino)-3-((tert-butoxycarbonyl)amino)propanoic acid
Formula:
C₁₆H₂₂N₂O₆ Storage Temperature: Refrigerator |
MDL Number:
MFCD00237342 CAS Number: 16947-84-5 |
Specification Test Results
Optical Rotation, [a]D²⁵ | –12.0 ±1° (C=1 in MeOH) |
¹³C NMR | Conforms to Structure |
¹H NMR | Conforms to Structure |
Purity by HPLC | ≥99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...