Sodium dihydrogen phosphate ≥99.0% for HPLC, Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic
17844-250G
17844-50G
BJ17844-250GEA
765
USD
BJ17844-250G
BJ17844-50G
Sodium dihydrogen phosphate ≥99.0% for HPLC, Fluka™
Sodium dihydrogen phosphate
Formula:
NaH₂PO₄ MW: 119.98 g/mol Melting Pt: 650 °C Density: 2.36 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003527 CAS Number: 7558-80-7 EINECS: 231-449-2 REACH: 01-2119489796-13 Merck Index: 12,08806 |
Specification Test Results
Assay | 99.0 - 101.0 % |
Appearance of the solution (c=10%) | complying |
pH (10 %, 20°C) | 4.0 - 4.5 |
Absorbance at 210 nm | Max. 0.1 |
Absorbance at 260 nm | Max. 0.06 |
Absorbance 280 nm | Max. 0.04 |
Absorbance at 320 nm | Max. 0.02 |
Absorbance 500 nm | Max. 0.02 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...