MutaPLATE® HLA DQ2+8 (TM) PCR Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
KF190532 KF190596
102993-576EA 1484.23 USD
102993-576 102993-578
MutaPLATE® HLA DQ2+8 (TM) PCR Assay Kit, Eagle Biosciences
Assays qPCR Assays
MutaPLATE® HLA-DQ2+8 (TM) real time PCR kit is a molecular biological test for detection of the genetic profile determining the HLA class II serotypes DQ2 and DQ8 (MHC system) (LPH) in open real time PCR systems.
  • For research use only
  • Sample type: DNA
MutaPLATE® HLA-DQ2+8 (TM) real time PCR Kit contains three allele specific primers and TaqMan probes, labeled with FAM, for the detection of the HLA-DQ2 and DQ8 alleles. The TaqMan probes bind to the amplificated target-DNA. Due to this, a fluorescence signal is generated and detected by the optical unit of the real time PCR instrument. Positive samples are identified due to the increase in fluorescence on the FAM (530 nm) channel during the PCR. An internal amplification control (IC), labeled with yakima yellow (560 nm), is also amplified with every sample. For a valid result, there must be a successful amplification of the IC on the YAK channel.
Precaución: Research use only and is not intended for diagnostic or therapeutic procedures.