LT250 Protective Eyewear, HexArmor
Supplier: HexArmor
11-18001-02 11-18002-02
76207-824EA 35.08 USD
76207-824 76207-826
LT250 Protective Eyewear, HexArmor
Safety Glasses
Safety eyewear should never be treated as one size fits all. Simply wearing any generic pair of safety eyewear isn’t enough to protect the eyes.
  • Fits over most prescription glasses
  • Hinged side arms fold flat for easy storage
  • Adaptive soft-grip side arms for a comfortable and secure fit
  • Contoured nose bridge for comfort and fit
  • Unrestricted peripheral vision with side protection
  • LTX™ Lens Technology for premium optical clarity
  • 100% UV protection
The LT250 with permanent anti-fog coatings. Featuring over-the-glasses (OTG) maximum protection, unrestricted peripheral vision, and high comfort safety.
Although it may decrease the possibility of injury, poorly or incorrectly fitting eyewear will put users at a higher risk for injury. HexArmor’s German-engineered dual-injection mold technology, allows the combination of soft and hard components into flex frames, making them comfortable and durable, yet highly adaptive.
They fit to most face widths or shapes, making wearability, protection, and compliance an easier endeavor.Soft earpieces limit ear pressure.
Certificaciones: Meets Standards ANSI Z87.1.2015, CE EN 166, CSA 94.3.