Gentamicin sulphate reagent_solution 10 mg/ml

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

091676045 0916760J8
10221-872EA 33.83 USD
10221-872 IC0916760J8
Gentamicin sulphate reagent_solution 10 mg/ml
Gentamicin sulphate

Gentamicin is an aminoglycoside antibiotic complex produced by fermentation of Micromonospora purpurea or M. echinospora. It is a mixture of 3 major components designated as C1 (C21H43N5O7), C1a (C19H39N5O), and C2 (C20H41N5O7). Gentamicin is used as the sulfate salt. Each component consists of five basic nitrogens and requires five equivalents of sulfuric acid per mole of gentamicin base. Gentamicin sulfate is a broad spectrum antibiotic. It inhibits the growth of a wide variety of Gram-positive and Gram-negative microorganisms, including strains resistant to tetracycline, chloramphenicol, kanamycin, and colistin, particularly strains of Pseudomonas, Proteus, Staphylococcus, and Streptococcus. It is effective against mycoplasma as well.


  • Concentration : 10 mg/mL
  • Potency : ≥590 µg/mg (dry basis), ≥483.8 µg/mg (as is)
  • Suitability : Suitable for cell culture
  • Product Type : Biochemicals
  • Chemical Class : Antibiotics
  • Presentation : Clear Colorless Solution
  • Format: Solution
  • pH : 4.0 - 6.0
  • NOTES : Osmolality: 29 mM/kg (Lit.)
  • Sterility : Sterile

Storage Temperature: Refrigerator
MDL Number: MFCD00270181
CAS Number: 1405-41-0
Merck Index: 13,04403
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=314cc7461af1dbdad64bfbe7b30c3be87aa2fc00&vid=937d04509a03d426dbebd2f1e93017daec49030d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Concentration 9.5 - 10.5 mg/ml
pH 4.0 - 6.0
Potency (dry basis) >590 µg/mg
Potency (as is) >483.8 µg/mg
Sterility Passes
Bacterial Inhibition Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR