G418 disulfate solution 50 mg/mL
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger
|
091672546
091672548
IC091672546EA
196.19
USD
IC091672546
IC091672548
G418 disulfate solution 50 mg/mL
G418 disulphate
G418 Sulfate is an aminoglycoside related to Gentamicin and is toxic to bacteria, yeast,higher plants, mammalian cells, protozoans and helminths. The resistance genes are located on transposons Tn601(903) and Tn5 and are bacterial in derivation, but they can be expressed in eukaryotic cells. By introducing these genes into cells, resistance to G418 Sulfate is conferred. G418 Sulfate is an effective selecting agent utilized in killing prokaryotic and eukaryotic cells.
- Application Areas: Eukaryotic Selection/Stable Cell Line Generation
- Optical Rotation: +112° (Lit.)
- UV/Visible Absorbance: A570 nm: 0.01, A280 nm: 0.001 (Lit.)
- Presentation: Clear, Colorless Solution
- pH: 5.3 (Lit.)
- Sterility: Sterile
- Sterilization of Solutions: Filtered through 0.22 µm micron filter
Formula:
C₂₀H₄₄N₄O₁₈S₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00058314 CAS Number: 108321-42-2 |
Specification Test Results
Identity Test | Passes |
Concentration (active G418 sulfate) | 50 ± 1 mg/ml |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...